Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium
GSA_VA

Catalog No. R01202 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
150/Pk.
20/Pk.
Format:
10mm x 100mm Spacesaver Plate
12mm x 85mm Monoplate
15mm x 103mm Tube
85mm Monoplate
Tube
Includes:
TSA w/10% Sheep Blood
TSA w/5% Sheep Blood
Manufacturing Origin:
United States
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Includes Manufacturing Origin
R01202 100/Pk. 85mm Monoplate TSA w/5% Sheep Blood United States
R01200 10/Pk. 85mm Monoplate TSA w/5% Sheep Blood United States
R060312 20/Pk. Tube TSA w/5% Sheep Blood United States
R01204 10/Pk. 12mm x 85mm Monoplate TSA w/10% Sheep Blood United States
R01198 150/Pk. 10mm x 100mm Spacesaver Plate TSA w/5% Sheep Blood United States
R060310 100/Pk. 15mm x 103mm Tube TSA w/5% Sheep Blood United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. R01202 Supplier Thermo Scientific™ Supplier No. R01202
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote hemolysis.

Grow a wide variety of fastidious organisms on Thermo Scientific™ Blood Agar (TSA with Sheep Blood), a general purpose non-selective medium with the addition of sheep blood in the formulation to promote the presence and visualization of haemolytic reactions so it can be used as a diagnostic tool.

Tryptone Soya Agar (casein soya bean digest agar) with defibrinated sheep blood is suitable for the cultivation of fastidious aerobic and anaerobic bacteria, the latter of which can be grown either in deep cultures or by incubation under anaerobic conditions. The combination of casein digest and papaic digest of soybean meal leads to optimal growth due to synergy between the protein supply of casein and the carbohydrate supply of soybeans. Sterile defibrinated sheep blood used to enrich the medium produces the hemolysis characteristics of different bacteria, such as streptococci, listeria, hemolytic Staphylococcus, Escherichia coli and Pseudomonas.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM.
Color Opaque red
Content And Storage 2°C to 8°C
Format 85mm Monoplate
Manufacturing Origin United States
Media Type Blood Agar
Product Type Agar Plate
Fill Volume 18.5 mL
Includes TSA w/5% Sheep Blood
Packaging Type Flow Wrapped - Single Layer
Quantity 100/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.