Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blood Agar/ Blood Agar (TSA w/5% Sheep Blood) Biplate 10Pk

Catalog No. R02020 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R02020 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R02020 Supplier Thermo Scientific™ Supplier No. R02020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grow a wide variety of microorganisms and detect, enumerate and differentiate the members of coliform group with this convenient bi-plate.

Grow a wide variety of microorganisms and detect, enumerate and differentiate the members of coliform group Thermo Scientific™ Remel™ Blood Agar/ Blood Agar (TSA w/5% Sheep Blood) Biplate.

Blood Agar (TSA w/5% Sheep Blood) is suitable for the cultivation of fastidious aerobic and anaerobic bacteria, the latter of which can be grown in either deep cultures or by incubation under anaerobic conditions.

Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with this convenient bi-plate.

  • Blood Agar (TSA w/5% Sheep Blood) facilitates rapid production of large colonies, clearly defined zones of hemolysis and good colonial differentiation.
  • Escherichia coli (purple colonies with a green metallic sheen) are easy to differentiate from Enterobacteria aerogenes (purple mucoid colonies).
  • Ready-to-use convenience of prepared media.
  • Space-saving bi-plate format.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Carmine red opaque
Content And Storage 2°C to 8°C
Description Blood Agar/ Blood Agar (TSA w/5% Sheep Blood) Biplate
Format Biplate
Manufacturing Origin United States
Product Type Agar Plate
Fill Weight 17g + 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.