Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blood Agar (TSA w/5% Sheep Blood)/EMB Agar (Levine) Biplate
GSA_VA

Catalog No. R02041 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R02041 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R02041 Supplier Thermo Scientific™ Supplier No. R02041
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grow a wide variety of microorganisms and detect, enumerate and differentiate the members of coliform group with this convenient bi-plate.

Grow a wide variety of microorganisms and detect, enumerate and differentiate the members of coliform group Thermo Scientific™ Remel™ Blood Agar (TSA w/5% Sheep Blood)/EMB Agar (Levine) Biplate.

Blood Agar (TSA w/5% Sheep Blood) is suitable for the cultivation of fastidious aerobic and anaerobic bacteria, the latter of which can be grown in either deep cultures or by incubation under anaerobic conditions. While EMB Agar (Levine) is a versatile medium used for the differentiation of Escherichia coli and Enterobacteria aerogenes and also for rapid identification of Candida albicans.

Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with this convenient bi-plate.

  • Blood Agar (TSA w/5% Sheep Blood) facilitates rapid production of large colonies, clearly defined zones of hemolysis and good colonial differentiation.
  • EMB Agar (Levine) is recommended for the detection, enumeration, and differentiation of members of the coliform group by American Public health Association.
  • EMB Agar (Levine) makes the results easy to read. Due to Eosin–Y and methylene blue indicators Escherichia coli (purple colonies with a green metallic sheen) are easy to differentiate from Enterobacteria aerogenes (purple mucoid colonies).
  • Ready-to-use convenience of prepared media.
  • Space-saving bi-plate format.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Carmine, red opaque/ Pearl ruby red, transparent
Content And Storage 2°C to 8°C
Description Blood Agar (TSA w/5% Sheep Blood)/ EMB Agar, Levine Biplate
Format Biplate
Manufacturing Origin United States
Product Type Agar Plate
Fill Weight 17g + 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.