Learn More
Med Vet International Blood Bag w/ ACD Solution, Equine, 3000ml, Sterile, J0520V, Each
Catalog No. 5000582293
Encompass_Preferred
The standard blood transfusion in equine medicine is not a common procedure. The large volumes needed to be effective can be a challenge to collect and administer. This is compounded by the lack of equine specific transfusion materials to make the process easier and simpler to do. Traditional equine blood collection / transfusion requires a large collection bag, then the separate addition of anticoagulants at collection time. JorVet now offers large collection bags with ACD anticoagulant already provided in the bag plus an integral attached 14g needle in two bag sizes. In addition, equine plasma can have a life saving role in newborn foals in conditions such as the"failure of passive antibody transfer" and neonatal isoerytholysis.3000ml sterile blood bag w/ 450ml ACD, 100cm tubing and 14g needle
Catalog No. 50-005-82293 Supplier Med Vet International Supplier No. JOR520V
The standard blood transfusion in equine medicine is not a common procedure. The large volumes needed to be effective can be a challenge to collect and administer. This is compounded by the lack of equine specific transfusion materials to make the process easier and simpler to do. Traditional equine blood collection / transfusion requires a large collection bag, then the separate addition of anticoagulants at collection time. JorVet now offers large collection bags with ACD anticoagulant already provided in the bag plus an integral attached 14g needle in two bag sizes. In addition, equine plasma can have a life saving role in newborn foals in conditions such as the"failure of passive antibody transfer" and neonatal isoerytholysis.3000ml sterile blood bag w/ 450ml ACD, 100cm tubing and 14g needle