Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Blood Bank Crossmatch Holders
SCI_ED

Catalog No. p-7146071 Shop All Fisherbrand Products
Change view
Click to view available options
Positions:
24
29
38
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S29187 24
S29188 29
S29189 38
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S29187
Only null left
Add to Cart
Add to Cart

For typing, screening, and other blood bank applications.

  • Clear acrylic shelves with white bases and sides
  • Holds donor sample tubes as well as tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening
  • Available in four sizes

24-tube capacity:
Holds four 16 mm O.D. sample tubes and twenty 10 mm O.D. crossmatch tubes

29-tube capacity:
Holds one 16 mm sample tube, one 13 mm pilot tube serum, and twenty-seven 10 mm phenotype, genotype, and cross-match tubes

38-tube capacity:
Holds thirty-eight tubes for five sets of tubes with printed identification on top panel

  • Set 1: One 16 mm patient, one serum tube, and one cell suspension tube
  • Set 2: Four 10 mm-14 mm forwarding group drives
  • Set 3: Three reverse grouping tubes
  • Set 4: Sixteen major cross match tubes
  • Set 5: Twelve minor cross match tubes

90-tube capacity:
Holds six 16 mm O.D. donor sample tubes, eighty-four 10 to 13 mm tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening

Specifications

Autoclavable Not Autoclavable
Color Clear, Transparent, White
Holds Tubes (10 and 16 mm)
Material Acrylic
Positions 24
For Use With (Equipment) Blood bank crossmatch tubes
Dimensions (L x W x H) 11.5 x 2.5 x 2.5 in. (29.21 x 6.35 x 6.35 cm)
Length (Metric) 29.21 cm
Length (English) 11.5 in.
Width (English) 2.5 in.
Height (English) 2.5 in.
Diameter (Metric) Opening 10 mm and 16 mm
Product Type Tube Rack
Height (Metric) 6.35 cm
Width (Metric) 6.35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.