Learn More
Medicus Health Blood Bank Transport Hardside Cooler, Medium 19 Qt
SDP
Catalog No. NC3736269
Encompass
Blood Bank Transport Cooler, 19 Quart can hold up several units of blood or other specimens as well as three ice packs. Cooler has been validated to hold temperature for at least 8 hours under normal hospital conditions. External Dimensions: 16.6"L x 11"W x 13"H.
Catalog No. NC3736269 Supplier Medicus Health Supplier No. 5319M5
Blood Bank Transport Cooler, 19 Quart can hold up several units of blood or other specimens as well as three ice packs. Cooler has been validated to hold temperature for at least 8 hours under normal hospital conditions. External Dimensions: 16.6"L x 11"W x 13"H.