Learn More
Sigma Aldrich Fine Chemicals Biosciences Blood-Brain Barrier hCMEC/D3 Cell Line The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function. |
Catalog No. NC2093100
Encompass
Blood-Brain Barrier hCMEC/D3 Cell Line The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function. |
Catalog No. NC2093100 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCC066
Blood-Brain Barrier hCMEC/D3 Cell Line The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function. |