Learn More
RNA Medical BLOOD COL TUBES SAFEWRAP 100PK
SDP
Catalog No. NC9223573
Encompass
Blood Collection Tube, Safe-Wrap Tubes are Heparinized Glass Capillary Devices Wrapped in a Puncture-Resistant Film, Used for Collection of Capillary Blood Samples for Measurement of pH, Blood Gases, Electrolytes, 220uL Maximum Fill
Catalog No. NC9223573 Supplier RNA Medical Supplier No. CT 220
Blood Collection Tube, Safe-Wrap Tubes are Heparinized Glass Capillary Devices Wrapped in a Puncture-Resistant Film, Used for Collection of Capillary Blood Samples for Measurement of pH, Blood Gases, Electrolytes, 220uL Maximum Fill