Learn More
Med Vet International Jorvet 450ml Blood Collection Unit, with 16g Needle, J0520
Catalog No. 502522330
Encompass_Preferred
CPDA-1 is the anticoagulant of choice in human blood banking. After collection, stored blood will have a 35-day shelf life. Bag comes with 16g diamond point non-detachable needle plus double spike ports. 450ml.
Catalog No. 50-252-2330 Supplier Med Vet International Supplier No. JOR520
CPDA-1 is the anticoagulant of choice in human blood banking. After collection, stored blood will have a 35-day shelf life. Bag comes with 16g diamond point non-detachable needle plus double spike ports. 450ml.