Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSP BUCA Blood Collection Accessory (BUCA)

Catalog No. 21888004
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-888-004 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-888-004 Supplier NSP BUCA Supplier No. BU50101
Add to Cart
Add to Cart
This item is not returnable. View return policy

A single use, sterile, and non-invasive device used for connecting to venous access device for the collection of blood specimen.

  • Single use device comprising of a polypropylene plastic holder, and an adapter with a male luer feature
  • A stainless steel needle covered with a rubber sleeve is attached to the adapter and is housed in the holder of the BUCA device
  • Needle stick issues are prevented due to a holder that encase the needle
  • The sample collection tube is also housed within the holder to protect the user or healthcare personnel from direct contact with blood specimens
  • Adapter allows connection to an interface device that provides collection of blood specimen into the blood collection tube that is inserted into the hollow of the holder of the BUCA device

Specifications

Disposable Single-use
No. of Places 2
Latex Free Yes
Product Type Blood Collection Accessory (BUCA)
Length (English) 15.75 in.
Length (Metric) 0.40 m
Storage Requirements 4°C to 32°C
Material Polypropylene
Autoclavable Not Autoclavable
For Use With (Equipment) Venous Access Devices
Sterility Sterile
Certifications/Compliance ISO 13485
Color Blue
For Use With (Application) Collection of Blood Specimen
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.