Learn More
BD Blood Collection Set
Catalog No. BD367326 Shop All Waters Corporation Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
BD367326 Sterile
1 options
This item is not returnable. View return policy
Catalog No. BD367326 Supplier BD Supplier No. 367326
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Intended for blood collection and short-term infusion
Push button without luer adapter button ensures irreversible needle lock green 19mm, tubing length 305mm 21G gauge BD Vacutainer
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blood Collection Set
Length (Metric) Needle 19.05 mm
Sterility Sterile
For Use With (Application) Intended for blood collection and short-term infusion
Latex Free Yes