Learn More
G-Biosciences Blood Collection Tube
SCI_ED
Catalog No. S04223
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
S04223 50/Pk.
1 options
Catalog No. S04223 Supplier G-Biosciences Supplier No. 7862183
Add to Cart
Add to Cart
Blood collection tube is used to collect and transport whole blood at ambient temperatures.
Blood collection tube is used to collect and transport whole blood at ambient temperatures. It collects an exact amount of whole blood while stabilizes DNA, RNA and inactivating any pathogens in the sample.
Specifications
Product Type Blood Collection Tube
Quantity 50/Pk.