Learn More
Sol-M™ Blood Collection Tube Holder with Luer Lock Adapter
Description
- Designed for collecting specimens from IV lines or for collecting urine specimens
- Compatible with IV sets, ports, and tubing according to ANSI standards
Specifications
Specifications
| Material | Polypropylene |
| Autoclavable | Not Autoclavable |
| Product Type | Standard Tube Holder |
| Disposable | Single-use |
| Latex Free | Yes |
| Sterility | Sterile |