Learn More
Sol-M™ Blood Collection Tube Holder
Description
- Compatible with standard multi-sample needles and blood collection tubes
Specifications
Specifications
| Material | Polypropylene |
| Autoclavable | Not Autoclavable |
| Product Type | Standard Tube Holder |
| Disposable | Single-use |
| For Use With (Equipment) | Butterfly Needles and Multi-sample Needles |
| Latex Free | Yes |
| Sterility | Sterile |