Learn More
Abnova™ Blood DNA Purification Kit (Pre-filled)
Catalog No. 89339132 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-339-132 1 Kit
1 options
Catalog No. 89-339-132 Supplier Abnova™ Supplier No. KA5510
Add to Cart
Add to Cart
Blood DNA Purification Kit (Pre-filled) is designed specifically for the optimal automated extraction of DNA from whole blood, frozen blood or buffy coat on the SpiralPipet™ instrument.
Specifications
Content And Storage Components under room temperature (15∼35 °C) can be stored until the expiration date labeled on the box. The Proteinase K was transported at room temperature. When received, please store at /20 °C. Repeating of freezing and thawing may cause the activity decay of Proteinase K.
Format Kit
Isolation Technology Magnetic Bead
Product Type DNA Purification Kit
Quantity 1 Kit