Learn More
Omnimed Inc BLOOD FILTRATION POWER LIFTER
SDP
Catalog No. NC3628236
Encompass_Preferred
Omnimed blood filtration power lifter stands are designed to provide hazard-free, ergonomic handling of blood collection materials. The single column system is constructed of anodized aluminum poles, a heavy duty cast aluminum base with 6 non-conductive casters and a wheel ring handle for easy transport. The powder coated 18 hanger assembly holds blood collection bags and leak depletion Filters. A Poly tray situated above the base contains 6 removable sections with 3 containers each to collect the filtered product. A total of 18 bags can be held in the tray. The pole assembly is raised or lowered with the use of foot pedals that operate the Pneumatic air glide system on the stand. The stands are simple to use and have an internal rechargeable 12V battery with the recharger, for cord free use. The adjustable height of the pole is 5' 7" to 9' from floor to top of the hanger. A low voltage light kit is an optional accessory for this unit.
Catalog No. NC3628236 Supplier Omnimed Inc Supplier No. 741318
Omnimed blood filtration power lifter stands are designed to provide hazard-free, ergonomic handling of blood collection materials. The single column system is constructed of anodized aluminum poles, a heavy duty cast aluminum base with 6 non-conductive casters and a wheel ring handle for easy transport. The powder coated 18 hanger assembly holds blood collection bags and leak depletion Filters. A Poly tray situated above the base contains 6 removable sections with 3 containers each to collect the filtered product. A total of 18 bags can be held in the tray. The pole assembly is raised or lowered with the use of foot pedals that operate the Pneumatic air glide system on the stand. The stands are simple to use and have an internal rechargeable 12V battery with the recharger, for cord free use. The adjustable height of the pole is 5' 7" to 9' from floor to top of the hanger. A low voltage light kit is an optional accessory for this unit.