Learn More
Mopec Blood Fixative
SDP
Catalog No. 22039020 Shop All Mopec Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
22-039-020 500 mL
1 options
Catalog No. 22-039-020 Supplier Mopec Supplier No. FS754
Add to Cart
Add to Cart
It helps maintain the clarity and integrity of blood-based evidence for accurate analysis.
  • Performance
    • Stabilizes blood-based prints and impressions
    • Prevents smearing, distortion, and detail loss
    • Prepares evidence for water-based enhancement methods
    • Preserves clarity and contrast during processing
    • Effective on porous and absorbent surfaces
  • Capacity
    • 500 mL
Specifications
Product Type Blood Fixative
Quantity 500 mL