Learn More
Norgen Biotek Corp Blood DNA Isolation Mini Kit, 50 Prep
Catalog No. NC1541532
Encompass_Preferred
Norgen’s Blood DNA Isolation Mini Kit is designed for the rapid preparation of DNA from up to 200 µL of whole blood using a rapid spin column protocol. DNA can be isolated from the blood of various species, including humans. Preparation time for a single sample is less than 30 minutes and each kit contains sufficient materials for 50 preparations. The kit isolates genomic DNA, mitochondrial DNA, viral DNA and bacterial DNA from blood samples. The purified DNA is of excellent quality and yield and completely compatible with downstream applications including PCR, qPCR, Sequencing, Genotyping, Virus Detection, Southern Blot analysis.
Catalog No. NC1541532 Supplier Norgen Biotek Corp Supplier No. 46300
Norgen’s Blood DNA Isolation Mini Kit is designed for the rapid preparation of DNA from up to 200 µL of whole blood using a rapid spin column protocol. DNA can be isolated from the blood of various species, including humans. Preparation time for a single sample is less than 30 minutes and each kit contains sufficient materials for 50 preparations. The kit isolates genomic DNA, mitochondrial DNA, viral DNA and bacterial DNA from blood samples. The purified DNA is of excellent quality and yield and completely compatible with downstream applications including PCR, qPCR, Sequencing, Genotyping, Virus Detection, Southern Blot analysis.