Learn More
Bioworld Blood Genomic DNA Isolation (Micro) Kit, 50 Prep(s)
Supplier:  Bioworld 107600991
Encompass_Preferred
For the rapid preparation of high quality DNA from whole blood, buffy coat, plasma and serum Input volumes up to 150 µL No phenol required for DNA isolation Extract DNA from whole blood, buffy coat, plasma and serum Fast and convenient spin column protocol FEATURES AND BENEFITSNo Phenol-Chloroform extraction or alcohol precipitation - Isolate genomic DNA without the use of harmful chemicals Fast and easy processing -Rapid spin-column format allows for the processing of multiple samples in 30 minutes. Isolate genomic DNA from various inputs - Isolate DNA from various blood inputs including plasma, whole blood, buffy coat and serum Recovered genomic DNA is suitable for downstream applications - Purified genomic DNA is fully compatible with restriction enzyme digestions, Southern Blot and PCR analysis. High quality DNA with no RNA contamination - No contamination or degradation of genomic DNA isolated with the Blood Genomic DNA Isolation Micro Kit is observed.
Catalog No. 50-196-956