Learn More
Bioworld Blood Genomic DNA Isolation Maxi, 12 Prep(s)
Catalog No. 50196976
Encompass_Preferred
For the rapid preparation of high quality DNA from whole blood, plasma and serum Input volumes of 2 mL to 10 mL No phenol required for DNA isolation Extract DNA from whole blood, plasma and serum Fast and convenient spin column protocol FEATURES AND BENEFITSNo Phenol-Chloroform extraction or alcohol precipitation - Isolate genomic DNA without the use of harmful chemicals. Fast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 30 minutes. Isolate genomic DNA from various inputs - Isolate DNA from various blood inputs including plasma, whole blood and serum Recovered genomic DNA is suitable for downstream applications - Purified genomic DNA is fully compatible with restriction enzyme digestions, Southern Blot and PCR analysis. High quality DNA with no RNA contamination - No contamination or degradation of genomic DNA isolated with the Blood Genomic DNA Isolation Maxi Kit is observed.
Catalog No. 50-196-976 Supplier Bioworld Supplier No. 107601201
For the rapid preparation of high quality DNA from whole blood, plasma and serum Input volumes of 2 mL to 10 mL No phenol required for DNA isolation Extract DNA from whole blood, plasma and serum Fast and convenient spin column protocol FEATURES AND BENEFITSNo Phenol-Chloroform extraction or alcohol precipitation - Isolate genomic DNA without the use of harmful chemicals. Fast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 30 minutes. Isolate genomic DNA from various inputs - Isolate DNA from various blood inputs including plasma, whole blood and serum Recovered genomic DNA is suitable for downstream applications - Purified genomic DNA is fully compatible with restriction enzyme digestions, Southern Blot and PCR analysis. High quality DNA with no RNA contamination - No contamination or degradation of genomic DNA isolated with the Blood Genomic DNA Isolation Maxi Kit is observed.