Learn More
Bioworld Blood Genomic DNA Isolation Mini, 50 Prep(s)
Catalog No. 50196974
Encompass_Preferred
For the rapid preparation of high quality DNA from whole blood, plasma and serum Input volumes of 200 µL to 500 µL No phenol required for DNA isolation Extract DNA from whole blood, plasma and serum Fast and convenient spin column protocol FEATURES AND BENEFITSNo Phenol-Chloroform extraction or alcohol precipitation - Isolate genomic DNA without the use of harmful chemicals Fast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 30 minutes Isolate genomic DNA from various inputs - Isolate DNA from various blood inputs including plasma, whole blood and serum Recovered genomic DNA is suitable for downstream applications - Purified genomic DNA is fully compatible with restriction enzyme digestions, Southern Blot and PCR analysis. High quality DNA with no RNA contamination - No contamination or degradation of genomic DNA isolated with the Blood Genomic DNA Isolation Mini Kit is observed.
Catalog No. 50-196-974 Supplier Bioworld Supplier No. 107601181
For the rapid preparation of high quality DNA from whole blood, plasma and serum Input volumes of 200 µL to 500 µL No phenol required for DNA isolation Extract DNA from whole blood, plasma and serum Fast and convenient spin column protocol FEATURES AND BENEFITSNo Phenol-Chloroform extraction or alcohol precipitation - Isolate genomic DNA without the use of harmful chemicals Fast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 30 minutes Isolate genomic DNA from various inputs - Isolate DNA from various blood inputs including plasma, whole blood and serum Recovered genomic DNA is suitable for downstream applications - Purified genomic DNA is fully compatible with restriction enzyme digestions, Southern Blot and PCR analysis. High quality DNA with no RNA contamination - No contamination or degradation of genomic DNA isolated with the Blood Genomic DNA Isolation Mini Kit is observed.