Learn More
Biochain Institute Inc vamPure Blood Nucleic Acid Extraction Kit, 1 kit/pk
SDP
Catalog No. 501871538
Encompass_Preferred
BioChain's vamPure Blood Nucleic Acid Extraction kit allows for fast and efficient genomic DNA extraction from whole blood samples. The DNA is eluted in EDTA-free buffer, allowing for immediate use in experiments where EDTA is not tolerated. EDTA may then be added separately for ideal long-term storage of the DNA. The magnetic bead-based extraction protocol is also ideally suited for automation using open systems. DNA extracted using this kit is suitable for PCR, qPCR, next generation sequencing (NGS), and other applications.
Catalog No. 50-187-1538 Supplier Biochain Institute Inc Supplier No. K5011720
BioChain's vamPure Blood Nucleic Acid Extraction kit allows for fast and efficient genomic DNA extraction from whole blood samples. The DNA is eluted in EDTA-free buffer, allowing for immediate use in experiments where EDTA is not tolerated. EDTA may then be added separately for ideal long-term storage of the DNA. The magnetic bead-based extraction protocol is also ideally suited for automation using open systems. DNA extracted using this kit is suitable for PCR, qPCR, next generation sequencing (NGS), and other applications.