Learn More
WELCH ALLYN INC BLOOD PRESSURE CUFF THIGH
Catalog No. NC9075371
Encompass
FlexiPort Reusable Blood Pressure Cuffs with One-Tube Tri-Purpose Connectors - 1 each Thigh [size 13] •Easy one-hand attachment/detachment of tubing with FlexiPort makes changing cuffs much faster and easier •Latex-free material minimizes risk of allergic reactions •Rotatable port reduces stress to cuff tubing and port, improving patient comfort and cuff durability •Folded edge reduces risk of cuts and scrapes, maximizing patient comfort •Meets all latest clinical guidelines for proper fit from AAMI and AHA
Catalog No. NC9075371 Supplier WELCH ALLYN INC Supplier No. REUSE131SC
FlexiPort Reusable Blood Pressure Cuffs with One-Tube Tri-Purpose Connectors - 1 each Thigh [size 13] •Easy one-hand attachment/detachment of tubing with FlexiPort makes changing cuffs much faster and easier •Latex-free material minimizes risk of allergic reactions •Rotatable port reduces stress to cuff tubing and port, improving patient comfort and cuff durability •Folded edge reduces risk of cuts and scrapes, maximizing patient comfort •Meets all latest clinical guidelines for proper fit from AAMI and AHA