Learn More
World Precision Instrument Blood Pressure Measurement for Rodents-II-MRBP-R
SDP
Catalog No. 502538754
Encompass
This revolutionary design brings non-invasive blood pressure testing to a new level — a true turn-key system for accurate, consistent blood pressure measurement on mice, rats or any other laboratory animal test subject.
It is a compact, simple yet versatile system that can test from one to 200 animals at a time with independent control of each channel. Simple daisy-chaining allows expansion of up to 200 independently controlled systems.
All components are built into one small unit — controls, inflation of tail cuff, warming environment with whisper-quiet fans — providing an ideal system for teaching facilities and for the pharmaceutical industry when high throughput is a must.
Catalog No. 50-253-8754 Supplier World Precision Instrument Supplier No. IIMRBPR
Add to Cart
Add to Cart
Request a Quote
This revolutionary design brings non-invasive blood pressure testing to a new level — a true turn-key system for accurate, consistent blood pressure measurement on mice, rats or any other laboratory animal test subject.
It is a compact, simple yet versatile system that can test from one to 200 animals at a time with independent control of each channel. Simple daisy-chaining allows expansion of up to 200 independently controlled systems.
All components are built into one small unit — controls, inflation of tail cuff, warming environment with whisper-quiet fans — providing an ideal system for teaching facilities and for the pharmaceutical industry when high throughput is a must.