Learn More
World Precision Instrument Blood Pressure Monitor
SDP
Catalog No. 502521048
Encompass
Audible monitor with variable pitch provides feedback
BP1 accepts WPI’s BLPR2 blood pressure transducer (right) as well as other blood pressure transducers. An audio monitor provides a signal with variable pitch and amplitude, allowing you to hear changes in blood pressure. Digital LCD display pro­vides average or peak signal values from 0 to 1999 mV. With an optional pressure gauge (not provided — see PM015D/R), you may cal­i­brate the display to read mmHg. Record­er out­put con­nec­tor al­lows di­rect con­nec­tion to a pen re­cord­er, os­cil­lo­scope or com­put­er via a data acquisition system.
Catalog No. 50-252-1048 Supplier World Precision Instrument Supplier No. SYSBP1
Audible monitor with variable pitch provides feedback
BP1 accepts WPI’s BLPR2 blood pressure transducer (right) as well as other blood pressure transducers. An audio monitor provides a signal with variable pitch and amplitude, allowing you to hear changes in blood pressure. Digital LCD display pro­vides average or peak signal values from 0 to 1999 mV. With an optional pressure gauge (not provided — see PM015D/R), you may cal­i­brate the display to read mmHg. Record­er out­put con­nec­tor al­lows di­rect con­nec­tion to a pen re­cord­er, os­cil­lo­scope or com­put­er via a data acquisition system.