Learn More
World Precision Instrument Blood Pressure Transducer & Cable
SDP
Catalog No. NC0957225
Encompass
BLPR2 can be used for the direct arterial and venous pressure measurement in animal blood vessels. Supplied sterile, BLPR2 is accurate, linear and stable with temperature. May be sterilized cold with Cidex or a similar bactericide.
Catalog No. NC0957225 Supplier World Precision Instrument Supplier No. BLPR2
BLPR2 can be used for the direct arterial and venous pressure measurement in animal blood vessels. Supplied sterile, BLPR2 is accurate, linear and stable with temperature. May be sterilized cold with Cidex or a similar bactericide.