Learn More
Alkali Scientific ASI™ Blood Separation Tube, PP, Conical, Sterile, 50mL, 300/Cs
SDP
Catalog No. 503026619
Encompass_Preferred
ASI™ Blood Separation Tube is a product that facilitates the separation of peripheral blood mononuclear cell (PBMC) from the blood. The filter inside the tube prevents mixing of the PBMC, lymphocyte and pellets. • Simplify injection of Separation media or blood samples • Fast separation • Prevent the pellets from mixing after removal ▪ Non-cytotoxic▪ Non-pyrogenic▪ DNase / RNase – free▪ Human DNA – free
Catalog No. 50-302-6619 Supplier Alkali Scientific Supplier No. BST50
ASI™ Blood Separation Tube is a product that facilitates the separation of peripheral blood mononuclear cell (PBMC) from the blood. The filter inside the tube prevents mixing of the PBMC, lymphocyte and pellets. • Simplify injection of Separation media or blood samples • Fast separation • Prevent the pellets from mixing after removal ▪ Non-cytotoxic▪ Non-pyrogenic▪ DNase / RNase – free▪ Human DNA – free