Learn More
Genesis Bps LLC BLOOD TEMP 10 INDICATOR 100PK
Catalog No. NC0842329
Encompass_Preferred
The Blood Temp 10 blood bag indicator is designed for use by blood banks and transfusion services during blood storage and transportation. With the push of a button, Blood Temp 10 is activated and indicates simply and clearly if the core temperature of the blood has risen to 10 deg. C (50 deg. F) or above. Sold 100 per pack.
Catalog No. NC0842329 Supplier Genesis Bps LLC Supplier No. 3BT10
The Blood Temp 10 blood bag indicator is designed for use by blood banks and transfusion services during blood storage and transportation. With the push of a button, Blood Temp 10 is activated and indicates simply and clearly if the core temperature of the blood has risen to 10 deg. C (50 deg. F) or above. Sold 100 per pack.