Learn More
Enzo Life Sciences Blood and tissue DNA methylation kit (200tests)
SDP
Catalog No. 502012566
Encompass_Preferred
The Blood and tissue DNA methylation kit features simple and reliable DNA bisulfite conversion directly from blood, tissue, and cells without the prerequisite for DNA purification. The sensitivity of this kit makes it possible to amplify bisulfite converted DNA from as few as 10 cells or 50pg DNA. Desulphonation and clean-up of the converted DNA is performed using a unique low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing, etc.
Catalog No. 50-201-2566 Supplier Enzo Life Sciences Supplier No. ENZ450040200
The Blood and tissue DNA methylation kit features simple and reliable DNA bisulfite conversion directly from blood, tissue, and cells without the prerequisite for DNA purification. The sensitivity of this kit makes it possible to amplify bisulfite converted DNA from as few as 10 cells or 50pg DNA. Desulphonation and clean-up of the converted DNA is performed using a unique low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing, etc.