Learn More
Med Vet International Blood Transfer Unit 300ml, J0520TB , Each
Catalog No. 5000582315
Encompass_Preferred
Designed to receive blood components or plasma, transfer bags have 2 spike entry ports and a single transfer spike with tubing. Sterile. Do not contain anticoagulants. 300mL.
Catalog No. 50-005-82315 Supplier Med Vet International Supplier No. JOR520TB
Designed to receive blood components or plasma, transfer bags have 2 spike entry ports and a single transfer spike with tubing. Sterile. Do not contain anticoagulants. 300mL.