Learn More
Mopec Blood Tube Protector (Various Sizes)
SDP
Catalog No. 22446343 Shop All Mopec Products
Change view
Click to view available options
Includes:
Double Tube
Single Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-446-343 Double Tube
22-446-342 Single Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-343 Supplier Mopec Supplier No. FS358
Add to Cart
Add to Cart
Designed to safeguard blood collection tubes during handling, transport, and storage, these durable, impact-resistant protectors help prevent breakage and leakage, reducing the risk of sample loss or contamination and supporting secure specimen handling.
Construction
  • Self-sealing bubble pack
  • Protects blood tubes in shipping or storage
  • FS357: (1) compartment
  • FS358: (2) compartments to prevent the glass tubes from touching each other
Quantity
  • 250 per case
Options and Accessories
  • Available as protector for single or double tubes
  • Note: Tubes sold separately
Specifications
Includes Double Tube
Quantity 250/Cs.