Learn More
Medicus Health Blood Tube Rocker
SDP
Catalog No. NC0139535
Encompass
∙ Tube Rocker holds up to 15 test tubes and silently rocks them at a fixed rate of 25 RPM for gentle mixing ∙Blood Tube Rocker perfect for hematology departments and small physician offices∙Now with new integrated "pause" button which allows you to stop the "rocking" temporarily in order to retrieve tubes without burning up the motor∙25 RPM rocking motion preserves integrity of fragile blood cells∙Protects samples from being hemolyzed or traumatized∙Great for mixing control samples before use∙Perfect for slide preparation areas to use before making a smear when Diff is indicated∙The reversible non-slip pad is autoclavable and will accommodate larger and hard to mix tubes∙Holds 15 tubes up to 120mm long∙Compact size uses minimal bench space∙Overall Dimensions 13.7"W x 4.7"D x 5"H
Catalog No. NC0139535 Supplier Medicus Health Supplier No. 5265M2
∙ Tube Rocker holds up to 15 test tubes and silently rocks them at a fixed rate of 25 RPM for gentle mixing ∙Blood Tube Rocker perfect for hematology departments and small physician offices∙Now with new integrated "pause" button which allows you to stop the "rocking" temporarily in order to retrieve tubes without burning up the motor∙25 RPM rocking motion preserves integrity of fragile blood cells∙Protects samples from being hemolyzed or traumatized∙Great for mixing control samples before use∙Perfect for slide preparation areas to use before making a smear when Diff is indicated∙The reversible non-slip pad is autoclavable and will accommodate larger and hard to mix tubes∙Holds 15 tubes up to 120mm long∙Compact size uses minimal bench space∙Overall Dimensions 13.7"W x 4.7"D x 5"H