Learn More
Description
- Five milliliter vial contains enough antisera for 75 students
- Sample will keep over one year with proper refrigeration
- Materials from human-derived cell lines were found to be negative in tests for Hepatitis B, HIV and EBV
- Includes Anti-A and Anti-B, Anti Rh and Anti-H Lectin Anti-sera
DOT: Non regulated
Specifications
Specifications
| Content And Storage | Ambient |
| Type | Blood Typing Anti-Sera Type A, B and Rh |
| For Use With (Application) | For Blood typing of real Human Blood |
| Includes | Anti-A and Anti-B and Anti Rh Anti-sera |
| Packaging Type | Plastic Bottle |
| Quantity | 1 Ea. |
For educational use only. Not FDA approved for clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
