Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Blood Typing Kits with Simulated Blood
GREENER_CHOICESCI_ED

Catalog No. S96792 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Blood Type Kit
Innovating Science Refill
For Use With (Application):
Simulated abo blood typing
Simulated abo/rh blood typing
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
S96792 Blood Type Kit Simulated abo/rh blood typing
S96793 Blood Type Kit Simulated abo blood typing
S96793R Innovating Science Refill Simulated abo blood typing
S96792R Innovating Science Refill Simulated abo/rh blood typing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S96792 Supplier Innovating Science™ Supplier No. IS3101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Four simulated blood samples
  • One bottle each of simulated anti-A and anti-B serum
  • 40 blood typing trays
  • ABO/Rh kit also includes one bottle of simulated anti-Rh serum

Determine the ABO or ABO/Rh blood type of four unknown samples.

Use hands-on activities to understand the nature and importance of antigen-antibody reactions.

  • Provide accurate simulation of the blood typing procedure
  • No toothpicks, stirring or waiting for results
  • Kit contains materials for 10 groups

Kits use simulated blood only.

Order Info

Kit refills do not include blood typing trays.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13+ years
For Use With (Application) Simulated abo/rh blood typing
Includes 4 simulated blood samples, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1bottle simulated anti-rh serum, 40 blood typing trays, Teacher's manual and student study guide copymasters
Quantity 1
Class Size 10 Lab Groups
Product Type Blood Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.