Learn More
Medicus Health Blood Unit Boot Tray with Handle, 4 Place
SDP
Catalog No. NC2225518
Encompass
∙ Designed to store a maximum of 4 Blood Unit Boot Stands in the Storage Boot so they are clearly visible∙ Blood Unit Boot Tray with Handles are made from a clear PETG plastic that's built to last ∙ Don't settle for cheap acrylic knockoffs that crack easily, yellow over time and splinter when you try to clean or wipe them down with standard cleaning products∙ Blood Unit Boot Tray with Handle is an effective and efficient way to manage your blood inventory∙ Keeps blood upright and separated so Blood Bag label expiration dates, blood types and bar codes are clearly visible∙ Blood Unit Boot Tray has dividers to keep bags upright and separated by blood type or crossmatched units but dividers are also removable which allows more storage space if needed∙ Blood Unit Boot Tray has integral handles ∙ Dimensions: 5""""W x 9.5""""D x 4.5""""H
This item is not returnable. View return policy
Catalog No. NC2225518 Supplier Medicus Health Supplier No. 2597M2
∙ Designed to store a maximum of 4 Blood Unit Boot Stands in the Storage Boot so they are clearly visible∙ Blood Unit Boot Tray with Handles are made from a clear PETG plastic that's built to last ∙ Don't settle for cheap acrylic knockoffs that crack easily, yellow over time and splinter when you try to clean or wipe them down with standard cleaning products∙ Blood Unit Boot Tray with Handle is an effective and efficient way to manage your blood inventory∙ Keeps blood upright and separated so Blood Bag label expiration dates, blood types and bar codes are clearly visible∙ Blood Unit Boot Tray has dividers to keep bags upright and separated by blood type or crossmatched units but dividers are also removable which allows more storage space if needed∙ Blood Unit Boot Tray has integral handles ∙ Dimensions: 5""""W x 9.5""""D x 4.5""""H