Learn More
Medicus Health Blood Bank Cooler, Compact 7.5 Qt
SDP
Catalog No. NC2734288
Encompass
∙ Transport blood efficiently with this 7.5 quart blood bank transport cooler from Medicus Health ∙ Includes a prominent sticker to remind staff to return units to the Blood Bank∙ Great option for Blood Banks to send units using this blood cooler to the ER or Surgery to help prevent units from returning over temperature∙ Stainless Steel hardware provides many years of durable service∙ Convenient handle and heavy duty shoulder strap Constructed of durable polypropylene for easy cleaning∙ Insulated with High-Grade molded polystyrene poamnon-absorbent surface ∙ ∙ Offers superior protection for your blood products∙ Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙ Please note: Because this item is exposed to blood and other hazardous specimens it is non-returnable ∙ Internal Dimensions: 10.5"W x 7.25"D x 6.25"H; External Dimensions: 12.25"W x 8"D x 10"H
Catalog No. NC2734288 Supplier Medicus Health Supplier No. 5323M5
∙ Transport blood efficiently with this 7.5 quart blood bank transport cooler from Medicus Health ∙ Includes a prominent sticker to remind staff to return units to the Blood Bank∙ Great option for Blood Banks to send units using this blood cooler to the ER or Surgery to help prevent units from returning over temperature∙ Stainless Steel hardware provides many years of durable service∙ Convenient handle and heavy duty shoulder strap Constructed of durable polypropylene for easy cleaning∙ Insulated with High-Grade molded polystyrene poamnon-absorbent surface ∙ ∙ Offers superior protection for your blood products∙ Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙ Please note: Because this item is exposed to blood and other hazardous specimens it is non-returnable ∙ Internal Dimensions: 10.5"W x 7.25"D x 6.25"H; External Dimensions: 12.25"W x 8"D x 10"H