Learn More
STEMCELL Technologies BloodStor 100, Size: 50 mL
SDP
Catalog No. 501974442
Encompass
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor 100, an animal component-free and serum-free biopreservation medium. BloodStor 100 has been sterilized, tested for endotoxin and manufactured under cGMPs with USP-grade components. BloodStor 100 contains 100% (w/v) DMSO and is available in a variety of formats. Visit STEMCELL.com for more information.
Catalog No. 50-197-4442 Supplier STEMCELL Technologies Supplier No. 07951
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor 100, an animal component-free and serum-free biopreservation medium. BloodStor 100 has been sterilized, tested for endotoxin and manufactured under cGMPs with USP-grade components. BloodStor 100 contains 100% (w/v) DMSO and is available in a variety of formats. Visit STEMCELL.com for more information.