Learn More
STEMCELL Technologies BloodStor 55-5, Size: 115.2 mL
SDP
Catalog No. NC1751141
Encompass
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor 55-5, an animal component-free and ready-to-use medium. cGMP-manufactured with USP-grade components, BloodStor 55-5 is sterile and tested for cell-based release and endotoxins. BloodStor 55-5 is pre-formulated with 55% (w/v) DMSO USP, 5% (w/v) Dextran-40 USP, and WFI-quality water. It is available in a variety of formats. Visit STEMCELL.com for more information.
Catalog No. NC1751141 Supplier STEMCELL Technologies Supplier No. 07937
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor 55-5, an animal component-free and ready-to-use medium. cGMP-manufactured with USP-grade components, BloodStor 55-5 is sterile and tested for cell-based release and endotoxins. BloodStor 55-5 is pre-formulated with 55% (w/v) DMSO USP, 5% (w/v) Dextran-40 USP, and WFI-quality water. It is available in a variety of formats. Visit STEMCELL.com for more information.