Learn More
Epoch Life Science Inc GenCatch™ Blood &Tissue Genomic Miniprep Kit (50 preps) Uniquely formulated buffers, reinforced columns with collection tubes, along with protocols.
SDP
Catalog No. 501538093
Encompass_Preferred
Provides a simple, fast, cost-effective method to extract gDNA ready for downstream use from various starting materials (such as bacteria, yeast, blood, cells, tissue, feces, etc.). Silica membrane technology. 120 ug Binding capacity. Instructions for sample type listed in protocol. Used by labs around the world. 18 month shelf life.
Catalog No. 50-153-8093 Supplier Epoch Life Science Inc Supplier No. 2460050
Provides a simple, fast, cost-effective method to extract gDNA ready for downstream use from various starting materials (such as bacteria, yeast, blood, cells, tissue, feces, etc.). Silica membrane technology. 120 ug Binding capacity. Instructions for sample type listed in protocol. Used by labs around the world. 18 month shelf life.