Learn More
Agilent Technologies BLOS CERAMIC BEAD ASSEMBLY
Catalog No. NC3310736
Encompass
  • Enhanced selectivity for nitrogen and phosphorus compounds.
  • Preconditioned for guaranteed performance and reliability.
  • Self-aligning mechanism for easy and rapid installation.
  • Compatible with various Agilent GC systems.
  • Constructed from ceramic materials.
  • Includes NPD bead assembly, cap, and test chromatogram.
  • Conditioned as new.
  • Preconditioning process included.
Catalog No. NC3310736 Supplier Agilent Technologies Supplier No. G343460806
  • Enhanced selectivity for nitrogen and phosphorus compounds.
  • Preconditioned for guaranteed performance and reliability.
  • Self-aligning mechanism for easy and rapid installation.
  • Compatible with various Agilent GC systems.
  • Constructed from ceramic materials.
  • Includes NPD bead assembly, cap, and test chromatogram.
  • Conditioned as new.
  • Preconditioning process included.