Learn More
Research Products International Corp Blot Stripping Buffer 4X Solution, 250 ML
SDP
Catalog No. 501978314
Encompass_Preferred
Product Formulation: TRIS-HCl: 250 mM; SDS: 8%; pH: 6.8. Add mercaptoethanol according to protocol.
Instructions :
1. Dilute 4X Stripping Bufffer ( 1part Stripping Buffer: 3 parts distilled water) and add beta-mercaptoethanol (0.6%).
2. Wash the wet membrane in the stripping buffer for 5 minutes.
3. Incubate the membrane in stripping buffer for 30 minutes at 65 degrees Celius while agitating.
Catalog No. 50-197-8314 Supplier Research Products International Corp Supplier No. B41700250.0
Add to Cart
Add to Cart
Product Formulation: TRIS-HCl: 250 mM; SDS: 8%; pH: 6.8. Add mercaptoethanol according to protocol.
Instructions :
1. Dilute 4X Stripping Bufffer ( 1part Stripping Buffer: 3 parts distilled water) and add beta-mercaptoethanol (0.6%).
2. Wash the wet membrane in the stripping buffer for 5 minutes.
3. Incubate the membrane in stripping buffer for 30 minutes at 65 degrees Celius while agitating.