Learn More
SignaGen Laboratories BLOT STRIPPING REAGENT 125ML
Catalog No. 504773
Encompass
Western Blotting Reagents, Western Blotting Reagents, BlotFresh Western Blot Stripping Reagent was formulated for effective and convenient dissociation even very high affinity antibodies from Western blot without removing transferred proteins, thereby al
Catalog No. 50-477-3 Supplier SignaGen Laboratories Supplier No. SL100324S
Western Blotting Reagents, Western Blotting Reagents, BlotFresh Western Blot Stripping Reagent was formulated for effective and convenient dissociation even very high affinity antibodies from Western blot without removing transferred proteins, thereby al