Learn More
SignaGen Laboratories BLOT STRIPPING REAGENT 500ML
Catalog No. 504771
Encompass
Western Blotting Reagents, BlotFresh Plus Western Blot Stripping Reagent was developed for effectively removing hard-to-strip antibodies from blot for re-probing.
Catalog No. 50-477-1 Supplier SignaGen Laboratories Supplier No. SL100325
Western Blotting Reagents, BlotFresh Plus Western Blot Stripping Reagent was developed for effectively removing hard-to-strip antibodies from blot for re-probing.