Learn More
S BIO BlotGlyco Glycan Purification and Labeling Kit
Catalog No. NC3616335
Encompass
BlotGlyco is a sample preparation kit for glycan analysis of biological samples. Using this kit will purify and label glycans from the crude samples such as glycoprotein, serum, cell, organ, plant tissue and so on. The label is selectable depending on detection methods including HPLC, LC-MS, MALDI-TOF MS and so on. BlotGlyco beads are coated with dense hydrazide functional groups, which enable specific binding with aldehyde groups at the reducing end of glycan. The hydrazide groups on the beads form stable covalent bond with aldehyde; therefore, it allows the vigorous washing steps to eliminate peptide and other impurities.
Catalog No. NC3616335 Supplier S BIO Supplier No. BS45407Z
BlotGlyco is a sample preparation kit for glycan analysis of biological samples. Using this kit will purify and label glycans from the crude samples such as glycoprotein, serum, cell, organ, plant tissue and so on. The label is selectable depending on detection methods including HPLC, LC-MS, MALDI-TOF MS and so on. BlotGlyco beads are coated with dense hydrazide functional groups, which enable specific binding with aldehyde groups at the reducing end of glycan. The hydrazide groups on the beads form stable covalent bond with aldehyde; therefore, it allows the vigorous washing steps to eliminate peptide and other impurities.