Learn More
Sigma Aldrich Fine Chemicals Biosciences Blotting Nylon 66 membrane1EA
Catalog No. 502908659
Encompass
Blotting nylon provided on a roll is a standard item used in transfer of protein or nucleic acids during Western Southern and Northern procedures. This membrane is positively charged nylon cast on a polyester support. It comes as a roll sized 0.2m x 3m and can be cut to any size.
Catalog No. 50-290-8659 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 153561EA
Blotting nylon provided on a roll is a standard item used in transfer of protein or nucleic acids during Western Southern and Northern procedures. This membrane is positively charged nylon cast on a polyester support. It comes as a roll sized 0.2m x 3m and can be cut to any size.