Learn More
Bioworld Blotto/Antifoam in PBS, 500 mL
Catalog No. 50255457
Encompass_Preferred
Blotto/Antifoam in PBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk + 0.01% Antifoam A + 0.02% Sodium azide in PBS
Catalog No. 50-255-457 Supplier Bioworld Supplier No. 402200322
Blotto/Antifoam in PBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk + 0.01% Antifoam A + 0.02% Sodium azide in PBS