Learn More
Bioworld Blotto/Antifoam in PBS, 250 mL
Catalog No. 50255456
Encompass_Preferred
Blotto/Antifoam in PBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk + 0.01% Antifoam A + 0.02% Sodium azide in PBS
Catalog No. 50-255-456 Supplier Bioworld Supplier No. 402200321
Blotto/Antifoam in PBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk + 0.01% Antifoam A + 0.02% Sodium azide in PBS