Learn More
Bioworld Blotto/Antifoam in TBS, 500 mL
Catalog No. 50255459
Encompass_Preferred
Blotto/Antifoam in TBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.01% Anti-Foam A, 0.02% Sodium azide in TBS
Catalog No. 50-255-459 Supplier Bioworld Supplier No. 402200362
Blotto/Antifoam in TBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.01% Anti-Foam A, 0.02% Sodium azide in TBS