Learn More
Rockland Immunochemicals BLOTTO B PRE-MIXED - for a 1X SOLUTION of Immunoanalytical Grade 1% NON-FAT DRY MILK, 1% BSA in TBS with TWEEN-20 - B553-0500
SDP
Catalog No. 501058872
Encompass_Preferred
Rockland Immunochemicals produces the following blocking buffer reagents: BSA, BLOTTO, BLOTTO A, BLOTTO B and Blocking Buffer for Fluorescent Western Blotting (see related products below).  These products are intended to block non-specific binding of proteins in various immunological assays. Typically, western blotting and ELISA methods call for the addition of a blocking agent prior to and during incubation with primary antibody.
Catalog No. 50-105-8872 Supplier Rockland Immunochemicals Supplier No. B5530500
Rockland Immunochemicals produces the following blocking buffer reagents: BSA, BLOTTO, BLOTTO A, BLOTTO B and Blocking Buffer for Fluorescent Western Blotting (see related products below).  These products are intended to block non-specific binding of proteins in various immunological assays. Typically, western blotting and ELISA methods call for the addition of a blocking agent prior to and during incubation with primary antibody.