Learn More
Bioworld Blotto (5%) in PBS, 500 mL
Catalog No. 50255452
Encompass_Preferred
Blotto in PBS Blot blocking buffer used to block non-specific binding for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.02% Sodium azide in PBS
Catalog No. 50-255-452 Supplier Bioworld Supplier No. 402200241
Blotto in PBS Blot blocking buffer used to block non-specific binding for antibodies on the membrane Technical Information: 5% nonfat dry milk, 0.02% Sodium azide in PBS